73522

Sigma-Aldrich

Atto Rho101 maleimide

NACRES:
NA.32

fluorescence

λex 586 nm; λem 610 nm

Quality Level

storage temp.

−20°C

General description

Atto Rho101 maleimide is a new rhodamine dye, based on the well-known laser dye Rhodamine 101. The new label is functionalized for coupling to bio-molecules such as DNA, RNA or proteins. Atto Rho101 shows extraordinary brightness and photostability. After coupling to a substrate Atto Rho101 maleimide carries a net electrical charge of +1. The dye is moderately hydrophilic.

Legal Information

This product is for Research use only. In case of intended commercialization, please contact the IP-holder (ATTO-TEC GmbH, Germany) for licensing.

RIDADR

NONH for all modes of transport

WGK Germany

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service

Social Media

LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon

MilliporeSigma

Research. Development. Production.

We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.

© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.

Reproduction of any materials from the site is strictly forbidden without permission.