76140

Sigma-Aldrich

Atto Rho14 Biotin

Pricing and availability is not currently available.

fluorescence

λex 625 nm; λem 646 nm

storage temp.

−20°C

General description

Atto Rho14 is a rhodamine featuring a functionality for coupling to biomolecules such as DNA, RNA or proteins. The label shows strong absorption, and extraordinarily high fluorescence quantum yield. In fact Atto Rho14 is the
brightest label available in this wavelength range. In addition the dye exhibits an exceptionally high photostability.
After coupling to a substrate Atto Rho14 carries a net electrical charge of +1.
Absorption and fluorescence are pH-independent in the range of pH 2 to 11, used in typical applications. The dye is moderately hydrophilic.

find more information
here

Legal Information

This product is for Research use only. In case of intended commercialization, please contact the IP-holder (ATTO-TEC GmbH, Germany) for licensing.

RIDADR

NONH for all modes of transport

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service

Social Media

LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon

MilliporeSigma

Research. Development. Production.

We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.

© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.

Reproduction of any materials from the site is strictly forbidden without permission.