87785

Sigma-Aldrich

Atto Rho6G maleimide

NACRES:
NA.32
Pricing and availability is not currently available.

fluorescence

λex 535 nm; λem 560 nm

storage temp.

−20°C

General description

Atto Rho6G maleimide is a new rhodamine dye, based on the well-known laser dye Rhodamine 6G. The new label is functionalized for coupling to biomolecules such as DNA, RNA or proteins. Atto Rho6G maleimide shows strong absorption, extraordinary high fluorescence quantum yield, high thermal and photo-stability, and very little triplet formation.
After coupling to a substrate Atto Rho6G maleimide carries a net electric charge of +1.
The label is moderately hydrophilic

find more information
here

RIDADR

NONH for all modes of transport

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service

Social Media

LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon

MilliporeSigma

Research. Development. Production.

We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.

© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.

Reproduction of any materials from the site is strictly forbidden without permission.