93685

Sigma-Aldrich

Atto Rho3B

NACRES:
NA.32

Quality Level

fluorescence

λex 564 nm; λem 592 nm

storage temp.

−20°C

General description

Atto Rho3B is a new fluorescent label related to the well-known dye Rhodamin B. The dye shows strong absorption and high thermal and photo-stability. The characteristic feature of the dye is its temperature dependent fluorescence yield. The dye is moderately hydrophilic. Atto Rho3B is a cationic dye. After coupling to a substrate the dye carries a net electrical charge of +1.

find more information
here

RIDADR

NONH for all modes of transport

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service

Social Media

LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon

MilliporeSigma

Research. Development. Production.

We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.

© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.

Reproduction of any materials from the site is strictly forbidden without permission.