93688

Sigma-Aldrich

Atto 490LS azide

suitable for fluorescence

NACRES:
NA.32
Pricing and availability is not currently available.

mol wt

Mw 896 g/mol

fluorescence

λex 496 nm; λem 655.0 nm±10.0 nm in PBS, pH 7.4

suitability

suitable for fluorescence

storage temp.

−20°C

General description

Atto 490LS is a new fluorescent label featuring an extraordinary large Stokes-Shift of 165 nm. Thus the emission spectrum is almost completely separated from its absorption spectrum, making the dye highly suitable for multiplexing experiments, in particular in combination with Atto 488 and Atto 514. Atto 490LS is very hydrophillic and shows excellent water solubility. The dye exhibits a relatively high fluorescence quantum yield, which is only slightly reduced after conjugation to biomolecules, e.g. proteins, even at high degrees of labeling (DOL). Atto 490LS is an anionic dye. After conjugation to a substrate the dye carries a net electrical charge of -1.
The azide modification is used in the Huisgen reaction (“Click Chemistry“).

find more information
here

RIDADR

NONH for all modes of transport

WGK Germany

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Certificate of Analysis
Certificate of Origin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service

Social Media

LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon

MilliporeSigma

Research. Development. Production.

We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.

© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.

Reproduction of any materials from the site is strictly forbidden without permission.