All Photos(1)

V000317

Sigma-Aldrich

Sodium phosphate dibasic

p.a.

Synonym(s):
Sodium hydrogenphosphate, Disodium phosphate, Disodium hydrogen phosphate, sec-Sodium phosphate
Linear Formula:
Na2HPO4
CAS Number:
Molecular Weight:
141.96
EC Number:
MDL number:
PubChem Substance ID:

grade

p.a.

pKa (25 °C)

(1) 2.15, (2) 6.82, (3) 12.38 (phosphoric acid)

SMILES string

[Na+].[Na+].[H]OP([O-])([O-])=O

InChI

1S/2Na.H3O4P/c;;1-5(2,3)4/h;;(H3,1,2,3,4)/q2*+1;/p-2

InChI key

BNIILDVGGAEEIG-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

WGK Germany

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Certificate of Analysis

Certificate of Origin

Kaisorn L Chaichana et al.
Journal of neuroscience methods, 180(1), 116-125 (2009-05-12)
The discoveries of neural (NSCs) and brain tumor stem cells (BTSCs) in the adult human brain and in brain tumors, respectively, have led to a new era in neuroscience research. These cells represent novel approaches to studying normal phenomena such...
Janelle M Spinazzola et al.
Human molecular genetics, 24(9), 2470-2481 (2015-01-22)
Loss of gamma-sarcoglycan (γ-SG) induces muscle degeneration and signaling defects in response to mechanical load, and its absence is common to both Duchenne and limb girdle muscular dystrophies. Growing evidence suggests that aberrant signaling contributes to the disease pathology; however...
Natalie K Binder et al.
Molecular human reproduction, 21(5), 424-434 (2015-03-04)
The rate of obesity among men of reproductive age has tripled in the last three decades. Previously, we demonstrated that paternal obesity resulted in impaired preimplantation developmental kinetics, compromised post-compaction metabolism and decreased blastocyst cell number when embryos were generated...
Megan A Rees et al.
Journal of proteome research, 14(1), 120-132 (2014-10-21)
Interactions between a host and a bacterial pathogen are mediated by cross-talk between molecules present on, or secreted by, pathogens and host binding-molecules. Identifying proteins involved at this interface would provide substantial insights into this interaction. Although numerous studies have...
Filis Kazazi-Hyseni et al.
Pharmaceutical research, 31(10), 2844-2856 (2014-05-16)
The aim of this study was the development of poly(D,L-lactide-co-glycolide) (PLGA) microspheres with controlled porosity, to obtain microspheres that afford continuous release of a macromolecular model compound (blue dextran). PLGA microspheres with a size of around 40 μm and narrow size...

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service