All Photos(2)

V900130

Sigma-Aldrich

Sodium benzoate

Vetec, reagent grade, 98%

Synonym(s):
Benzoic acid sodium salt
Linear Formula:
C6H5COONa
CAS Number:
Molecular Weight:
144.10
Beilstein:
3572467
EC Number:
MDL number:
PubChem Substance ID:

grade

reagent grade

product line

Vetec

assay

98%

mp

>300 °C (lit.)

SMILES string

[Na+].[O-]C(=O)c1ccccc1

InChI

1S/C7H6O2.Na/c8-7(9)6-4-2-1-3-5-6;/h1-5H,(H,8,9);/q;+1/p-1

InChI key

WXMKPNITSTVMEF-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Packaging

500 g in poly bottle

Legal Information

Vetec is a trademark of Sigma-Aldrich International GmbH

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

13 - Non Combustible Solids

WGK Germany

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Certificate of Analysis

Certificate of Origin

Sandesh C S Nagamani et al.
Genetics in medicine : official journal of the American College of Medical Genetics, 14(5), 501-507 (2012-01-14)
The urea cycle consists of six consecutive enzymatic reactions that convert waste nitrogen into urea. Deficiencies of any of these enzymes of the cycle result in urea cycle disorders (UCDs), a group of inborn errors of hepatic metabolism that often...
Yue Mu et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 83(1), 130-135 (2011-09-06)
The toxicity of sodium benzoate to trypsin was investigated by fluorescence spectroscopy, synchronous fluorescence spectroscopy, UV-visible absorption spectroscopy and circular dichroism (CD) spectroscopy under mimic physiological conditions. Sodium benzoate could unfold trypsin by decreasing the β-sheet structure, which leads to...
Trelita de Sousa et al.
Bioresource technology, 123, 256-262 (2012-09-04)
Pseudomonas nitroreducens TSB.MJ10 exhibiting growth and bioemulsifier production with 0.5% sodium benzoate as the sole carbon source was isolated from a mangrove ecosystem in the vicinity of a petroleum pump. The bioemulsifier is a lipopeptide that is stable over a...
Jane Pui Ki Tsang et al.
Pediatric neurology, 47(4), 263-269 (2012-09-12)
In areas without expanded newborn screening, instead of presenting neonatally, patients with arginase deficiency typically present with spastic paraplegia in early childhood. Diagnosis of this rare neurometabolic disease poses the first challenge because it is often misdiagnosed as cerebral palsy...
Tony Jin et al.
Journal of food science, 76(3), M184-M188 (2011-05-04)
Survival of Escherichia coli O157:H7 and Salmonella Stanley on apples as affected by application of polylactic acid (PLA) coating with antimicrobials was investigated. Golden Delicious apples were spot inoculated with E. coli O157:H7 or S. Stanley and spray coated with PLA solutions containing lactic acid (LA), disodium...

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service