MilliporeSigma
All Photos(2)

V900374

Sigma-Aldrich

L-Ornithine monohydrochloride

Vetec, reagent grade, 98%

Synonym(s):
(S)-2,5-Diaminopentanoic acid monohydrochloride
Empirical Formula (Hill Notation):
C5H12N2O2 · HCl
CAS Number:
Molecular Weight:
168.62
Beilstein:
3625847
EC Number:
MDL number:
PubChem Substance ID:

grade

reagent grade

product line

Vetec

assay

98%

form

powder

color

white

SMILES string

Cl.NCCC[C@H](N)C(O)=O

InChI

1S/C5H12N2O2.ClH/c6-3-1-2-4(7)5(8)9;/h4H,1-3,6-7H2,(H,8,9);1H/t4-;/m0./s1

InChI key

GGTYBZJRPHEQDG-WCCKRBBISA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Packaging

100 g in poly bottle

Other Notes

Product of arginine degradation by arginase

Legal Information

Vetec is a trademark of Sigma-Aldrich International GmbH

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Certificate of Analysis

Enter Lot Number to search for Certificate of Analysis (COA).

Certificate of Origin

Enter Lot Number to search for Certificate of Origin (COO).

More Documents

SDS

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service