All Photos(1)

V900529

Sigma-Aldrich

Methyl phenylacetate

Vetec, reagent grade, 98%

Linear Formula:
C6H5CH2CO2CH3
CAS Number:
Molecular Weight:
150.17
Beilstein:
878795
EC Number:
MDL number:
PubChem Substance ID:

grade

reagent grade

product line

Vetec

assay

98%

refractive index

n20/D 1.503 (lit.)

bp

218 °C (lit.)

density

1.066 g/mL at 20 °C (lit.)

SMILES string

COC(=O)Cc1ccccc1

InChI

1S/C9H10O2/c1-11-9(10)7-8-5-3-2-4-6-8/h2-6H,7H2,1H3

InChI key

CRZQGDNQQAALAY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Packaging

250 g in glass bottle

Legal Information

Vetec is a trademark of Sigma-Aldrich International GmbH

Storage Class Code

10 - Combustible liquids

WGK Germany

WGK 2

Flash Point(F)

195.8 °F - closed cup

Flash Point(C)

91 °C - closed cup

Certificate of Analysis

Certificate of Origin

Miriam Frida Karlsson et al.
Journal of agricultural and food chemistry, 57(13), 5903-5909 (2009-06-06)
The Guatemalan moth Tecia solanivora is an invasive pest of potato in Central and South America. The larvae infest potato tubers in the field as well as in storage facilities. The headspace of potato foliage and potato tubers was studied...
Dengchao Li et al.
Preparative biochemistry & biotechnology, 43(2), 207-216 (2013-01-11)
This study developed a simple, efficient method for producing racemic β-phenylalanine acid (BPA) and its derivatives via the enantioselective acylation catalyzed by the penicillin G acylase from Alcaligenes faecalis (Af-PGA). When the reaction was run at 25°C and pH 10...
David M Suckling et al.
Tuberculosis (Edinburgh, Scotland), 91(4), 327-328 (2011-05-07)
The proboscis extension reflex in honeybees was evaluated for detection of tuberculosis. Restrained bees were tested with methyl phenylacetate, methyl p-anisate, and methyl nicotinate, previously identified from Mycobacterium tuberculosis cultures, to determine honeybee capacity for signature volatile detection. Methyl p-anisate...
Jose C S dos Santos et al.
Enzyme and microbial technology, 77, 1-7 (2015-07-04)
Lipase from Thermomyces lanuginosus (TLL) and lipase B from Candida antarctica (CALB) have been immobilized on divinylsulfone (DVS) activated agarose beads at pH 10 for 72 h. Then, as a reaction end point, very different nucleophiles have been used to...
Mona Syhre et al.
Tuberculosis (Edinburgh, Scotland), 88(4), 317-323 (2008-02-26)
Worldwide, tuberculosis (TB) kills nearly 2 million people annually, yet rapid diagnosis still relies on a 100-year-old method of sputum staining for acid-fast bacilli. The advent of solid phase micro-extraction and gas chromatography/mass spectrometry makes it possible to systematically investigate...

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service