V900798

Sigma-Aldrich

Acetic acid

Vetec, reagent grade, ≥99%

Synonym(s):
Glacial acetic acid
Linear Formula:
CH3CO2H
CAS Number:
Molecular Weight:
60.05
Beilstein/REAXYS Number:
506007
EC Number:
MDL number:
PubChem Substance ID:

grade

reagent grade

vapor density

2.07 (vs air)

product line

Vetec

assay

≥99%

form

liquid

autoignition temp.

800 °F

expl. lim.

16 %, 92 °F
4 %, 59 °F

refractive index

n20/D 1.371 (lit.)

pH

2.5 (20 °C, 50  g/L)

bp

117-118 °C (lit.)

mp

16.2 °C (lit.)

density

1.049 g/mL at 25 °C (lit.)

SMILES string

CC(O)=O

InChI

1S/C2H4O2/c1-2(3)4/h1H3,(H,3,4)

InChI key

QTBSBXVTEAMEQO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Packaging

500 mL in glass bottle

Legal Information

Vetec is a trademark of Sigma-Aldrich International GmbH

Pictograms

FlameCorrosion

Signal Word

Danger

Hazard Statements

hazcat

Eye Dam. 1 - Flam. Liq. 3 - Skin Corr. 1A

storage_class_code

3 - Flammable liquids

WGK Germany

WGK 1

Flash Point(F)

102.2 °F

Flash Point(C)

39 °C

Certificate of Analysis

Certificate of Origin

Katharine Trenholme et al.
Antimicrobial agents and chemotherapy, 58(7), 3666-3678 (2014-04-16)
Therapies to prevent transmission of malaria parasites to the mosquito vector are a vital part of the global malaria elimination agenda. Primaquine is currently the only drug with such activity; however, its use is limited by side effects. The development...
Takashi Taniguchi et al.
Arthritis & rheumatology (Hoboken, N.J.), 67(2), 517-526 (2014-11-12)
Fli-1, a potential predisposing factor for systemic sclerosis (SSc), is constitutively down-regulated in the lesional skin of patients with SSc by an epigenetic mechanism. To investigate the impact of Fli-1 deficiency on the induction of an SSc phenotype in various...
Isabelle Brunet et al.
The Journal of clinical investigation, 124(7), 3230-3240 (2014-06-18)
Autonomic sympathetic nerves innervate peripheral resistance arteries, thereby regulating vascular tone and controlling blood supply to organs. Despite the fundamental importance of blood flow control, how sympathetic arterial innervation develops remains largely unknown. Here, we identified the axon guidance cue...
N Robichaud et al.
Oncogene, 34(16), 2032-2042 (2014-06-10)
The progression of cancers from primary tumors to invasive and metastatic stages accounts for the overwhelming majority of cancer deaths. Understanding the molecular events which promote metastasis is thus critical in the clinic. Translational control is emerging as an important...
Xianbao Deng et al.
The New phytologist, 201(4), 1469-1483 (2013-11-26)
• Chalcone synthase (CHS) is the key enzyme in the first committed step of the flavonoid biosynthetic pathway and catalyzes the stepwise condensation of 4-coumaroyl-CoA and malonyl-CoA to naringenin chalcone. In plants, CHS is often encoded by a small family...

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service