Sign In to View Organizational & Contract Pricing.
Select a Size
25 MG
$23.40
1 G
$27.30
100 MG
$50.70
5 G
$106.60
10 G
$159.90
25 G
$215.80
100 G
$3,559.40
Vendor SKU: AA0036G9-25MG
$23.40
Ships in 10 days - from Aldrich Partner.
About This Item
Skip To
Inchi Code
1S/C25H30N4O9S2.C7H8O3S/c1-24(2)17(29-20(32)16(21(29)39-24)27-19(31)15(26)12-8-6-5-7-9-12)22(33)37-11-38-23(34)18-25(3,4)40(35,36)14-10-13(30)28(14)18;1-6-2-4-7(5-3-6)11(8,9)10/h5-9,14-18,21H,10-11,26H2,1-4H3,(H,27,31);2-5H,1H3,(H,8,9,10)/t14-,15-,16-,17+,18+,21-;/m1./s1
Inchi Key
FFCSPKNZHGIDQM-CGAOXQFVSA-N
Purity
98%
Storage temp.
2-8C
Country of Origin
US
Looking for similar products? Visit Product Comparison Guide
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service