Skip to Content
MilliporeSigma
Get 22% off for Pi Day through March 31.Save Now

AABH93DE36B1

AA BLOCKS, INC.

Fluorescein isothiocyanate-dextran

Synonym(s):

((3'-hydroxy-3-oxo-3H-spiro[isobenzofuran-1,9'-xanthen]-6'-yl)oxy)carbonimidothioic O,S-acid

Sign In to View Organizational & Contract Pricing.

Select a Size

5 MG

$59.80

10 MG

$81.90

25 MG

$104.00

100 MG

$182.00

250 MG

$377.00

Vendor SKU: AA00EBER-5MG

$59.80


Ships in 10 days - from Aldrich Partner.


Skip To

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

Inchi Code

1S/C21H13NO6S/c23-11-5-7-15-17(9-11)26-18-10-12(28-22-20(25)29)6-8-16(18)21(15)14-4-2-1-3-13(14)19(24)27-21/h1-10,23H,(H2,22,25,29)

Inchi Key

PUQQBNGWUKSTLB-UHFFFAOYSA-N

Purity

95%

Storage temp.

2-8C

Country of Origin

US


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service