Skip to Content
MilliporeSigma

AABH93DE5B89

AA BLOCKS, INC.

Laminin (929-933)

Synonym(s):

(2S,5S,11S,14S)-14-amino-11-((S)-sec-butyl)-2-(3-((diaminomethylene)amino)propyl)-5-(hydroxymethyl)-15-(4-hydroxyphenyl)-4,7,10,13-tetraoxo-3,6,9,12-tetraazapentadecanoic acid

Sign In to View Organizational & Contract Pricing.

Select a Size

Product AABH93DE5B89 is currently not available for purchase. Contact Technical Service

Skip To

Purity

95%

Inchi Code

1S/C26H42N8O8/c1-3-14(2)21(34-22(38)17(27)11-15-6-8-16(36)9-7-15)24(40)31-12-20(37)32-19(13-35)23(39)33-18(25(41)42)5-4-10-30-26(28)29/h6-9,14,17-19,21,35-36H,3-5,10-13,27H2,1-2H3,(H,31,40)(H,32,37)(H,33,39)(H,34,38)(H,41,42)(H4,28,29,30)/t14-,17-,18-,19-,21-/m0/s1

Inchi Key

MWOGMBZGFFZBMK-LJZWMIMPSA-N

Storage temp.

-20C

Country of Origin

US


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library