Skip to Content
MilliporeSigma

AABH97CD73A5

AA BLOCKS, INC.

Potassium guaiacolsulfonate (hemihydrate)

Synonym(s):

potassium 3-hydroxy-2-methoxybenzenesulfonate 4-hydroxy-3-methoxybenzenesulfonate hydrate

Sign In to View Organizational & Contract Pricing.

Select a Size

Product AABH97CD73A5 is currently not available for purchase. Contact Technical Service

Skip To

Inchi Code

1S/2C7H8O5S.2K.H2O/c1-12-7-4-5(13(9,10)11)2-3-6(7)8;1-12-7-5(8)3-2-4-6(7)13(9,10)11;;;/h2*2-4,8H,1H3,(H,9,10,11);;;1H2/q;;2*+1;/p-2

Inchi Key

NSEBUQNBYJQIRZ-UHFFFAOYSA-L

Purity

95%

Storage temp.

2-8C

Country of Origin

US


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library