Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

AABH97CDB7F8

AA BLOCKS, INC.

Suc-Gly-Gly-Phe-pNA

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$102.70
25 MG
$206.70
100 MG
$526.50
250 MG
$975.00

Vendor SKU: AA005ZD3-5MG

$102.70


Ships in 10 days - from Aldrich Partner.


Select a Size

Change View
5 MG
$102.70
25 MG
$206.70
100 MG
$526.50
250 MG
$975.00

About This Item

CAS Number:
Molecular Weight:
499.48
MDL number:

Vendor SKU: AA005ZD3-5MG

$102.70


Ships in 10 days - from Aldrich Partner.

Iupac Name

4-({2-[(2-{[(1S)-1-benzyl-2-(4-nitroanilino)-2-oxoethyl]amino}-2-oxoethyl)amino]-2-oxoethyl}amino)-4-oxobutanoic acid

Inchi Code

1S/C23H25N5O8/c29-19(10-11-22(32)33)24-13-20(30)25-14-21(31)27-18(12-15-4-2-1-3-5-15)23(34)26-16-6-8-17(9-7-16)28(35)36/h1-9,18H,10-14H2,(H,24,29)(H,25,30)(H,26,34)(H,27,31)(H,32,33)/t18-/m0/s1

InChI key

CFYIUBWVKZQDOG-SFHVURJKSA-N

storage temp.

-20C

Purity

99%

Country of Origin

US

Product Link

https://www.aablocks.com/prod/68982-90-1

Synonym

ChymotrypsinSubstrateI,Colorimetric

MSDS

https://www.aablocks.com/downloadSds?cas=68982-90-1


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service