Skip to Content
MilliporeSigma

AABH97CE2A8F

AA BLOCKS, INC.

2,4,4-trimethylpentanoic acid

Sign In to View Organizational & Contract Pricing

Select a Size


About This Item

CAS Number:
Molecular Weight:
144.21
MDL number:

Iupac Name

2,4,4-trimethylpentanoic acid

Inchi Code

1S/C8H16O2/c1-6(7(9)10)5-8(2,3)4/h6H,5H2,1-4H3,(H,9,10)

InChI key

UWXFTQSKZMNLSA-UHFFFAOYSA-N

storage temp.

RT

Purity

95%

Country of Origin

CN

Product Link

https://www.aablocks.com/prod/3302-09-8

MSDS

https://www.aablocks.com/downloadSds?cas=3302-09-8


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library