Skip to Content
MilliporeSigma

AABH97CE7713

AA BLOCKS, INC.

Iopentol

Synonym(s):

5-[acetyl(2-hydroxy-3-methoxypropyl)amino]-N~1~,N~3~-bis(2,3-dihydroxypropyl)-2,4,6-triiodoisophthalamide

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C20H28O9N3I3
CAS Number:
Molecular Weight:
835.17
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C20H28I3N3O9/c1-9(29)26(5-12(32)8-35-2)18-16(22)13(19(33)24-3-10(30)6-27)15(21)14(17(18)23)20(34)25-4-11(31)7-28/h10-12,27-28,30-32H,3-8H2,1-2H3,(H,24,33)(H,25,34)

Inchi Key

IUNJANQVIJDFTQ-UHFFFAOYSA-N

Purity

98%

Storage temp.

2-8C

Country of Origin

US





Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library