Skip to Content
MilliporeSigma

AABH97D3C036

AA BLOCKS, INC.

Mono(tetramethylammonium) tribromide

Synonym(s):

2λ⁴-tribroman-2-id-2-yl; tetramethylazanium

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C4H12N1Br3
CAS Number:
Molecular Weight:
313.86
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C4H12N.Br3/c1-5(2,3)4;1-3-2/h1-4H3;/q+1;-1

Inchi Key

JFZBJMYWDMJRDB-UHFFFAOYSA-N

Purity

98%

Storage temp.

N/A

Country of Origin

CN





Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library