Skip to Content
MilliporeSigma

AABH9A21B042

AA BLOCKS, INC.

Decarboxy Fexofenadine

Synonym(s):

4-{4-[hydroxy(diphenyl)methyl]-1-piperidinyl}-1-(4-isopropylphenyl)-1-butanol

Sign In to View Organizational & Contract Pricing.

Select a Size

25 MG

$759.20

100 MG

$1,609.40

Vendor SKU: AA00ADNQ-25MG

$759.20


Ships in 21 days - from Aldrich Partner.


About This Item

Linear Formula:
C31H39O2N1
CAS Number:
Molecular Weight:
457.66
MDL number:

Skip To

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

Inchi Code

1S/C31H39NO2/c1-24(2)25-15-17-26(18-16-25)30(33)14-9-21-32-22-19-29(20-23-32)31(34,27-10-5-3-6-11-27)28-12-7-4-8-13-28/h3-8,10-13,15-18,24,29-30,33-34H,9,14,19-23H2,1-2H3

Inchi Key

FZSQPQGERLSIOR-UHFFFAOYSA-N

Purity

95%

Storage temp.

-20C

Country of Origin

US


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service