Skip to Content
MilliporeSigma

AABH9A953E2E

AA BLOCKS, INC.

2-amino-4-(1-adamantyl) Thiazole

Sign Into View Organizational & Contract Pricing

Select a Size


About This Item

CAS Number:
Molecular Weight:
234.37
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

Iupac Name

4-((3r,5r,7r)-adamantan-1-yl)thiazol-2-amine

Inchi Code

1S/C13H18N2S/c14-12-15-11(7-16-12)13-4-8-1-9(5-13)3-10(2-8)6-13/h7-10H,1-6H2,(H2,14,15)/t8-,9+,10-,13?

InChI key

JVJWJUQLRDIVJV-JKTANTLQSA-N

storage temp.

2~8 C

Purity

97%

Country of Origin

US

Product Link

https://www.aablocks.com/prod/19735-74-1

MSDS

https://www.aablocks.com/downloadSds?cas=19735-74-1


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service