Skip to Content
MilliporeSigma

AABH9A954577

AA BLOCKS, INC.

Lead(II) acetate trihydrate

Synonym(s):

acetic acid, lead salt hydrate

Sign In to View Organizational & Contract Pricing.

Select a Size

Product AABH9A954577 is currently not available for purchase. Contact Technical Service

Skip To

Inchi Code

1S/C2H4O2.H2O.Pb/c1-2(3)4;;/h1H3,(H,3,4);1H2;

Inchi Key

ATQIWGVXMGLXBT-UHFFFAOYSA-N

Purity

99.998%

Storage temp.

2-8C

Country of Origin

US


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library