Skip to Content
MilliporeSigma

AABH9A954AA6

AA BLOCKS, INC.

Poly(tetrafluoroethylene)

Slide 1 of 1
Sign Into View Organizational & Contract Pricing

Select a Size

Product AABH9A954AA6 is currently not available for purchase. Contact Technical Service

About This Item

CAS Number:
Molecular Weight:
117.05
MDL number:

Skip To

Marketplace Support
Need help or have questions about this product?
Contact Seller

Inchi Code

1S/C2F4.H3N/c3-1(4)2(5)6;/h;1H3

Inchi Key

BQASMSZYTHSWSE-UHFFFAOYSA-N

storage temp.

RT

Country of Origin

US

Product Link

https://www.aablocks.com/prod/9002-84-0

MSDS

https://www.aablocks.com/downloadSds?cas=9002-84-0


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library