Select a Size
$221.00
$568.10
Vendor SKU: AA009KQE-1MG
$221.00
Skip To
Inchi Code
1S/C53H86O22/c1-23-32(58)36(62)39(65)43(69-23)75-42-38(64)34(60)25(19-55)71-46(42)72-26-20-67-45(41(35(26)61)74-44-40(66)37(63)33(59)24(18-54)70-44)73-31-10-11-49(5)27(47(31,2)3)8-12-50(6)28(49)9-13-53-29-16-48(4,21-56)14-15-52(29,22-68-53)30(57)17-51(50,53)7/h21,23-46,54-55,57-66H,8-20,22H2,1-7H3/t23-,24+,25+,26-,27-,28+,29+,30+,31-,32-,33+,34+,35-,36+,37-,38-,39+,40+,41+,42+,43-,44-,45-,46-,48-,49-,50+,51-,52+,53-/m0/s1
Inchi Key
ZDIHSHLFPFGAGP-LLEYBADXSA-N
Purity
95%
Storage temp.
-20C
Country of Origin
US
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service