Sign In to View Organizational & Contract Pricing.
Select a Size
Product AABH9A956A26 is currently not available for purchase. Contact Technical Service
About This Item
Skip To
Inchi Code
1S/C15H24N2O/c18-15-6-3-5-14-11-8-12(10-17(14)15)13-4-1-2-7-16(13)9-11/h11-14H,1-10H2/t11-,12-,13-,14+/m0/s1
Inchi Key
JYIJIIVLEOETIQ-XDQVBPFNSA-N
Purity
98%
Storage temp.
2-8C
Country of Origin
US
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.