Sign In to View Organizational & Contract Pricing.
Select a Size
1 MG
$520.00
5 MG
$1,231.10
Vendor SKU: AA02AP3D-1MG
$520.00
Ships in 21 days - from Aldrich Partner.
About This Item
Skip To
Inchi Code
1S/C19H18O4/c1-9-8-23-18-11-4-6-12-10(5-7-13(20)19(12,2)3)15(11)17(22)16(21)14(9)18/h4,6,9H,5,7-8H2,1-3H3/t9-/m0/s1
Inchi Key
ABVRZQXICIZPLM-VIFPVBQESA-N
Country of Origin
US
Storage temp.
2-8C
Purity
98%
MSDS
https://www.aablocks.com/downloadSds?cas=2470587-69-8
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service