Skip to Content
MilliporeSigma

AABH9AC99875

AA BLOCKS, INC.

(S)-(-)-1,1'-Bi-2-naphthol

Synonym(s):

[1,1'-binaphthalene]-3,3'-diol

Sign In to View Organizational & Contract Pricing.

Select a Size


About This Item

CAS Number:
Molecular Weight:
286.33
MDL number:

Inchi Code

1S/C20H14O2/c21-15-9-13-5-1-3-7-17(13)19(11-15)20-12-16(22)10-14-6-2-4-8-18(14)20/h1-12,21-22H

Inchi Key

CKBLTMGPJAACEY-UHFFFAOYSA-N

Storage temp.

RT

Country of Origin

US

Purity

98%

MSDS

https://www.aablocks.com/downloadSds?cas=18531-99-2


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library