Skip to Content
MilliporeSigma

AABH9BD8BA97

AA BLOCKS, INC.

(3β,5α,6α)-Cholest-5-ene-25,26,26,26,27,27,27-d7-3,6-diol

Synonym(s):

(3S,5S,6S,8S,9S,10R,13R,14S,17R)-10,13-dimethyl-17-((R)-6-(methyl-d3)heptan-2-yl-6,7,7,7-d4)hexadecahydro-1H-cyclopenta[a]phenanthrene-3,6-diol

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C27H48O2
CAS Number:
Molecular Weight:
411.72
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C27H48O2/c1-17(2)7-6-8-18(3)21-9-10-22-20-16-25(29)24-15-19(28)11-13-27(24,5)23(20)12-14-26(21,22)4/h17-25,28-29H,6-16H2,1-5H3/t18-,19+,20+,21-,22+,23+,24-,25+,26-,27-/m1/s1/i1D3,2D3,17D

Inchi Key

PMWTYEQRXYIMND-DNWVLYPESA-N

Purity

98%

Country of Origin

US

Storage temp.

N/A





Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library