MilliporeSigma
All Photos(1)

AC5388701317

Accela ChemBio Inc.

Methyl 4-Morpholinepropionate

CAS Number:
Molecular Weight:
173.21
MDL number:

Iupac Name

methyl 3-(4-morpholinyl)propanoate

Inchi Code

1S/C8H15NO3/c1-11-8(10)2-3-9-4-6-12-7-5-9/h2-7H2,1H3

InChI key

YMSLLIGKMYXCPK-UHFFFAOYSA-N

storage temp.

+4?

Purity

98%

Country of Origin

CN

Shipping Temperature

RT

Physical Form

Colorless to yellow liquid

Product Link

http://www.accelachem.com/cn/search.html?keyword=SY013056&attname=GoodsCode

Pictograms

Signal Word

Warning

Precautionary statements

MSDS

http://pdf.accelachem.com/cn/msds/SY013056.pdf

Certificate of Analysis

Enter Lot Number to search for Certificate of Analysis (COA).

Certificate of Origin

Enter Lot Number to search for Certificate of Origin (COO).

More Documents

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service