MilliporeSigma
All Photos(1)

AC5H97ED216C

Accela ChemBio Inc.

5-Nitro-1-pentene

CAS Number:
Molecular Weight:
115.13
MDL number:

Iupac Name

5-nitro-1-pentene

Inchi Code

1S/C5H9NO2/c1-2-3-4-5-6(7)8/h2H,1,3-5H2

InChI key

NPFOLTHIKZSVPQ-UHFFFAOYSA-N

storage temp.

+4?

Purity

95%

Country of Origin

CN

Shipping Temperature

RT

Physical Form

Colorless to yellow liquid

Product Link

http://www.accelachem.com/cn/search.html?keyword=SY266521&attname=GoodsCode

MSDS

http://pdf.accelachem.com/cn/msds/SY266521.pdf

Certificate of Analysis

Enter Lot Number to search for Certificate of Analysis (COA).

Certificate of Origin

Enter Lot Number to search for Certificate of Origin (COO).

More Documents

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service