Skip to Content
MilliporeSigma

ADV465749262

AChemBlock

Rostafuroxin

Sign Into View Organizational & Contract Pricing

Select a Size


About This Item

CAS Number:
Molecular Weight:
374.52
MDL number:

Iupac Name

(3S,5R,8R,9S,10S,13S,14S,17S)-17-(furan-3-yl)-10,13-dimethylhexadecahydro-14H-cyclopenta[a]phenanthrene-3,14,17-triol

Inchi Code

1S/C23H34O4/c1-20-8-5-17(24)13-15(20)3-4-19-18(20)6-9-21(2)22(25,10-11-23(19,21)26)16-7-12-27-14-16/h7,12,14-15,17-19,24-26H,3-6,8-11,13H2,1-2H3/t15-,17+,18+,19-,20+,21-,22+,23+/m1/s1

InChI key

AEAPORIZZWBIEX-DTBDINHYSA-N

storage temp.

Store at 0-8 C

Purity

98%

Country of Origin

CN

Product Link

https://www.achemblock.com/10371-rostafuroxin.html

pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary statements

MSDS

https://www.achemblock.com/10371-rostafuroxin.html


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library