Skip to Content
MilliporeSigma

ADV465749404

AChemBlock

Tolvaptan

Sign Into View Organizational & Contract Pricing

Select a Size


About This Item

CAS Number:
Molecular Weight:
448.95
MDL number:
Marketplace Support
Need help or have questions about this product?
Contact Seller
Marketplace Support
Need help or have questions about this product?
Contact Seller

Iupac Name

N-(4-{[(5R)-7-chloro-5-hydroxy-2,3,4,5-tetrahydro-1H-1-benzazepin-1-yl]carbonyl}-3-methylphenyl)-2-methylbenzamide

Inchi Code

1S/C26H25ClN2O3/c1-16-6-3-4-7-20(16)25(31)28-19-10-11-21(17(2)14-19)26(32)29-13-5-8-24(30)22-15-18(27)9-12-23(22)29/h3-4,6-7,9-12,14-15,24,30H,5,8,13H2,1-2H3,(H,28,31)/t24-/m1/s1

InChI key

GYHCTFXIZSNGJT-XMMPIXPASA-N

storage temp.

Store at 0-8 C

Purity

98%

Country of Origin

CN

Product Link

https://www.achemblock.com/10530-tolvaptan.html

pictograms

Exclamation mark

Signal Word

Not a hazardous substance or mixture

Hazard Statements

MSDS

https://www.achemblock.com/10530-tolvaptan.html


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library