MilliporeSigma
All Photos(1)

ADVH7F380C79

AChemBlock

Ethyl (pentafluorobenzoyl)acetate

CAS Number:
Molecular Weight:
282.17
MDL number:

Iupac Name

ethyl 3-oxo-3-(2,3,4,5,6-pentafluorophenyl)propanoate

Inchi Code

1S/C11H7F5O3/c1-2-19-5(18)3-4(17)6-7(12)9(14)11(16)10(15)8(6)13/h2-3H2,1H3

InChI key

SFYYMUUQGSQVFT-UHFFFAOYSA-N

Purity

95%

Country of Origin

CN

Product Link

https://www.achemblock.com/ethyl-pentafluorobenzoyl-acetate.html

Synonym

ethyl 3-oxo-3-(perfluorophenyl)propanoate

MSDS

https://www.achemblock.com/ethyl-pentafluorobenzoyl-acetate.html

Certificate of Analysis

Enter Lot Number to search for Certificate of Analysis (COA).

Certificate of Origin

Enter Lot Number to search for Certificate of Origin (COO).

More Documents

Quotes and Ordering

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service