Documents

ADVH7F3833CD

AChemBlock

tert-butyldimethyl((tributylstannyl)ethynyl)silane

CAS Number:
Molecular Weight:
429.35
MDL number:

Iupac Name

tert-butyldimethyl((tributylstannyl)ethynyl)silane

Inchi Code

1S/C8H15Si.3C4H9.Sn/c1-7-9(5,6)8(2,3)4;3*1-3-4-2;/h2-6H3;3*1,3-4H2,2H3;

InChI key

RWJZSEBIYYIYHQ-UHFFFAOYSA-N

Purity

95%

Country of Origin

CN

Product Link

https://www.achemblock.com/tert-butyldimethyl-tributylstannyl-ethynyl-silane.html

Synonym

tert-butyldimethyl((tributylstannyl)ethynyl)silane

MSDS

https://www.achemblock.com/tert-butyldimethyl-tributylstannyl-ethynyl-silane.html

Certificate of Analysis

Certificate of Origin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service