Documents

ADVH7F3838DD

AChemBlock

tert-butyl cinnamyl carbonate

CAS Number:
Molecular Weight:
234.3
MDL number:

Iupac Name

tert-butyl cinnamyl carbonate

Inchi Code

1S/C14H18O3/c1-14(2,3)17-13(15)16-11-7-10-12-8-5-4-6-9-12/h4-10H,11H2,1-3H3/b10-7+

InChI key

UZQQCYKWZUXRCV-JXMROGBWSA-N

Purity

95%

Country of Origin

CN

Product Link

https://www.achemblock.com/tert-butyl-cinnamyl-carbonate.html

Synonym

tert-butyl cinnamyl carbonate

MSDS

https://www.achemblock.com/tert-butyl-cinnamyl-carbonate.html

Certificate of Analysis
Certificate of Origin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service

Social Media

LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon

MilliporeSigma

Research. Development. Production.

We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.

© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.

Reproduction of any materials from the site is strictly forbidden without permission.