MilliporeSigma
All Photos(1)

ADVH9454794E

AChemBlock

AZ191

CAS Number:
Molecular Weight:
429.52
MDL number:

Iupac Name

N-(2-methoxy-4-(4-methylpiperazin-1-yl)phenyl)-4-(1-methyl-1H-pyrrolo[2,3-c]pyridin-3-yl)pyrimidin-2-amine

Inchi Code

1S/C24H27N7O/c1-29-10-12-31(13-11-29)17-4-5-21(23(14-17)32-3)28-24-26-9-7-20(27-24)19-16-30(2)22-15-25-8-6-18(19)22/h4-9,14-16H,10-13H2,1-3H3,(H,26,27,28)

InChI key

ZYVXTMKTGDARKR-UHFFFAOYSA-N

Purity

98%

Country of Origin

CN

Product Link

https://www.achemblock.com/az191.html

Synonym

n-(2-methoxy-4-(4-methylpiperazin-1-yl)phenyl)-4-(1-methyl-1h-pyrrolo[2,3-c]pyridin-3-yl)pyrimidin-2-amine

MSDS

https://www.achemblock.com/az191.html

Certificate of Analysis

Enter Lot Number to search for Certificate of Analysis (COA).

Certificate of Origin

Enter Lot Number to search for Certificate of Origin (COO).

More Documents

Quotes and Ordering

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service