Documents

ADVH97AC6C33

AChemBlock

[4-(2-Fluorocyclopropyl)phenyl]methanol

CAS Number:
Molecular Weight:
166.2

Iupac Name

(4-(2-fluorocyclopropyl)phenyl)methanol

Inchi Code

1S/C10H11FO/c11-10-5-9(10)8-3-1-7(6-12)2-4-8/h1-4,9-10,12H,5-6H2

InChI key

WNEPVDCRSQIOGI-UHFFFAOYSA-N

Purity

95.00%

Country of Origin

CN

Product Link

https://www.achemblock.com/4-2-fluorocyclopropyl-phenyl-methanol.html

Synonym

(4-(2-fluorocyclopropyl)phenyl)methanol

MSDS

https://www.achemblock.com/4-2-fluorocyclopropyl-phenyl-methanol.html

Certificate of Analysis

Certificate of Origin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service