ADVH98DEA573
AChemBlock
methyl 4-(4-methoxy-3,3-dimethyl-but-1-ynyl)benzoate
Iupac Name
methyl 4-(4-methoxy-3,3-dimethylbut-1-yn-1-yl)benzoate
Inchi Code
1S/C15H18O3/c1-15(2,11-17-3)10-9-12-5-7-13(8-6-12)14(16)18-4/h5-8H,11H2,1-4H3
InChI key
FBOCVSINUYYOHW-UHFFFAOYSA-N
Purity
97%
Country of Origin
CN
Product Link
https://www.achemblock.com/x181978-methyl-4-4-methoxy-3-3-dimethyl-but-1-ynyl-benzoate.html
Signal Word
Warning
Hazard Statements
Precautionary statements
MSDS
https://www.achemblock.com/x181978-methyl-4-4-methoxy-3-3-dimethyl-but-1-ynyl-benzoate.html
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service