ADVH9A1C1FF2
AChemBlock
Ethionamide (Nicotinamide Impurity)
About This Item
Iupac Name
2-ethylpyridine-4-carbimidothioic acid--nicotinimidic acid (1/1)
Inchi Code
1S/C8H10N2S.C6H6N2O/c1-2-7-5-6(8(9)11)3-4-10-7;7-6(9)5-2-1-3-8-4-5/h3-5H,2H2,1H3,(H2,9,11);1-4H,(H2,7,9)
InChI key
FZBCCDPMEOPGGV-UHFFFAOYSA-N
storage temp.
Purity
Country of Origin
Product Link
Synonym
Signal Word
Warning
Hazard Statements
Precautionary statements
MSDS
https://www.achemblock.com/w167858-ethionamide-nicotinamide-impurity.html
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.