Skip to Content
MilliporeSigma

ADVH9A1C771D

AChemBlock

(3-chloro-4-fluorophenyl)(4-fluoro-4-((methyl(5-methylpyrimidin-2-yl)amino)methyl)piperidin-1-yl)methanone

Slide 1 of 1
Sign Into View Organizational & Contract Pricing

Select a Size

250 MG
$529.00
1 G
$1,316.75

Vendor SKU: X209483-250MG

$529.00


Ships in 14 days - from Aldrich Partner.


Skip To

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

Iupac Name

(3-chloro-4-fluorophenyl)(4-fluoro-4-((methyl(5-methylpyrimidin-2-yl)amino)methyl)piperidin-1-yl)methanone

Inchi Code

1S/C19H21ClF2N4O/c1-13-10-23-18(24-11-13)25(2)12-19(22)5-7-26(8-6-19)17(27)14-3-4-16(21)15(20)9-14/h3-4,9-11H,5-8,12H2,1-2H3

InChI key

UIOBNHCZEKNJGR-UHFFFAOYSA-N

Purity

97%

Country of Origin

CN

Product Link

https://www.achemblock.com/x209483-3-chloro-4-fluorophenyl-4-fluoro-4-methyl-5-methylpyrimidin-2-yl-amino-methyl-piperidin-1-yl-methanone.html

pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary statements

MSDS

https://www.achemblock.com/x209483-3-chloro-4-fluorophenyl-4-fluoro-4-methyl-5-methylpyrimidin-2-yl-amino-methyl-piperidin-1-yl-methanone.html


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service