Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

ADVH9A1C9F2E

AChemBlock

RS1-PDK1 inhibitor

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
380.9

Iupac Name

N-(6-chlorobenzo[d]thiazol-2-yl)benzo[b]thiophene-3-sulfonamide

Inchi Code

1S/C15H9ClN2O2S3/c16-9-5-6-11-13(7-9)22-15(17-11)18-23(19,20)14-8-21-12-4-2-1-3-10(12)14/h1-8H,(H,17,18)

InChI key

VLGMTYRMKMVUHJ-UHFFFAOYSA-N

storage temp.

Store at 0-8 C

Purity

0.97

Country of Origin

CN

Product Link

https://www.achemblock.com/y227509-rs1-pdk1-inhibitor.html

Synonym

N-(6-chlorobenzo[d]thiazol-2-yl)benzo[b]thiophene-3-sulfonamide

pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary statements

MSDS

https://www.achemblock.com/y227509-rs1-pdk1-inhibitor.html


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library