ADVH9A9896FE
AChemBlock
Dodecyl-D-glucopyranoside
Sign Into View Organizational & Contract Pricing
Select a Size
5 G
$258.75
25 G
$891.25
Vendor SKU: T98353-5G
$258.75
Ships in 14 days - from Aldrich Partner.
Skip To
Iupac Name
dodecyl D-glucopyranoside
Inchi Code
1S/C18H36O6/c1-2-3-4-5-6-7-8-9-10-11-12-23-18-17(22)16(21)15(20)14(13-19)24-18/h14-22H,2-13H2,1H3/t14-,15-,16+,17-,18?/m1/s1
InChI key
PYIDGJJWBIBVIA-IHAUNJBESA-N
storage temp.
Store at 0-8 C
Purity
97%
Country of Origin
CN
Product Link
https://www.achemblock.com/t98353-dodecyl-d-glucopyranoside.html
Signal Word
Warning
Hazard Statements
Precautionary statements
MSDS
https://www.achemblock.com/t98353-dodecyl-d-glucopyranoside.html
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service