ADVH9AC94CE6
AChemBlock
N-Boc-N',N''-di-Cbz-L-arginine
Sign Into View Organizational & Contract Pricing
Select a Size
Product ADVH9AC94CE6 is currently not available for purchase. Contact Technical Service
Skip To
Iupac Name
(S)-5-(3-((benzyloxy)(hydroxy)methylene)-1-((benzyloxy)carbonyl)guanidino)-2-((tert-butoxy(hydroxy)methylene)amino)pentanoic acid
Inchi Code
1S/C27H34N4O8/c1-27(2,3)39-25(35)29-21(22(32)33)15-10-16-31(26(36)38-18-20-13-8-5-9-14-20)23(28)30-24(34)37-17-19-11-6-4-7-12-19/h4-9,11-14,21H,10,15-18H2,1-3H3,(H,29,35)(H,32,33)(H2,28,30,34)/t21-/m0/s1
InChI key
FVWNVCDDYWITMG-NRFANRHFSA-N
storage temp.
Store at Room Tem.
Purity
95.00%
Country of Origin
CN
Product Link
https://www.achemblock.com/x192224-n-boc-n-n-di-cbz-l-arginine.html
Signal Word
Warning
Hazard Statements
Precautionary statements
MSDS
https://www.achemblock.com/x192224-n-boc-n-n-di-cbz-l-arginine.html
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
