Skip to Content
MilliporeSigma

ADVH9B9B3A6E

AChemBlock

Ac-DEVD-CHO

Synonym(s):

(4S,7S,10S,13S)-7-(2-carboxyethyl)-4-(carboxymethyl)-13-formyl-10-isopropyl-2,5,8,11-tetraoxo-3,6,9,12-tetraazapentadecan-15-oic acid

Sign In to View Organizational & Contract Pricing.

Select a Size

5 MG

$264.50

25 MG

$684.25

Vendor SKU: AD265831-5MG

$264.50


Ships in 10 days - from Aldrich Partner.


About This Item

Linear Formula:
C20H30O11N4
CAS Number:
Molecular Weight:
502.48
MDL number:

Skip To

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

Inchi Code

1S/C20H30N4O11/c1-9(2)17(20(35)22-11(8-25)6-15(29)30)24-18(33)12(4-5-14(27)28)23-19(34)13(7-16(31)32)21-10(3)26/h8-9,11-13,17H,4-7H2,1-3H3,(H,21,26)(H,22,35)(H,23,34)(H,24,33)(H,27,28)(H,29,30)(H,31,32)/t11-,12-,13-,17-/m0/s1

Inchi Key

UMBVAPCONCILTL-MRHIQRDNSA-N

Purity

98%

Storage temp.

Store at -20 C

Country of Origin

CN

Physical Form

White Powder

Looking for similar products? Visit Product Comparison Guide

hstatements

signalword

Warning

pictograms

Exclamation mark

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service