Skip to Content
MilliporeSigma

ADVH9B9B4373

AChemBlock

Nourseothricin sulfate

Synonym(s):

sulfuric acid--(2R,3R,4S,5R,6R)-5-((S)-3,6-diaminohexanamido)-4-hydroxy-6-(((3aS,7R,7aS)-7-hydroxy-4-oxo-3a,4,5,6,7,7a-hexahydro-1H-imidazo[4,5-c]pyridin-2-yl)amino)-2-(hydroxymethyl)tetrahydro-2H-pyran-3-yl carbamate--(2R,3R,4S,5R,6R)-5-((S)-3-amino-6-((S)-3-amino-6-((S)-3,6-diaminohexanamido)hexanamido)hexanamido)-4-hydroxy-6-(((3aS,7R,7aS)-7-hydroxy-4-oxo-3a,4,5,6,7,7a-hexahydro-1H-imidazo[4,5-c]pyridin-2-yl)amino)-2-(hydroxymethyl)tetrahydro-2H-pyran-3-yl carbamate (1/1/1)

Sign In to View Organizational & Contract Pricing.

Select a Size

50 MG

$235.75

100 MG

$379.50

Vendor SKU: Y226261-50MG

$235.75


Ships in 10 days - from Aldrich Partner.


Skip To

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

Inchi Code

1S/C31H58N12O10.C19H34N8O8.H2O4S/c32-7-1-4-15(33)10-20(46)37-8-2-5-16(34)11-21(47)38-9-3-6-17(35)12-22(48)40-25-26(49)27(53-30(36)51)19(14-44)52-29(25)43-31-41-23-18(45)13-39-28(50)24(23)42-31;20-3-1-2-7(21)4-10(30)24-13-14(31)15(35-18(22)33)9(6-28)34-17(13)27-19-25-11-8(29)5-23-16(32)12(11)26-19;1-5(2,3)4/h15-19,23-27,29,44-45,49H,1-14,32-35H2,(H2,36,51)(H,37,46)(H,38,47)(H,39,50)(H,40,48)(H2,41,42,43);7-9,11-15,17,28-29,31H,1-6,20-21H2,(H2,22,33)(H,23,32)(H,24,30)(H2,25,26,27);(H2,1,2,3,4)/t15-,16-,17-,18+,19+,23+,24-,25+,26-,27-,29+;7-,8+,9+,11+,12-,13+,14-,15-,17+;/m00./s1

Inchi Key

HEPYPTBGGNPPAP-SPCBHCMPSA-N

Storage temp.

Store at 0-8 C

Purity

97.00%

Country of Origin

CN

MSDS

https://www.achemblock.com/y226261-nourseothricin-sulfate.html

hstatements

signalword

Warning

pictograms

Exclamation mark

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service