Sign In to View Organizational & Contract Pricing
Select a Size
Product ADVH9B9B4739 is currently not available for purchase. Contact Technical Service
Skip To
Iupac Name
potassium 4-hydroxy-3-methoxybenzenesulfonate hydrate
Inchi Code
1S/2C7H8O5S.2K.H2O/c2*1-12-7-4-5(13(9,10)11)2-3-6(7)8;;;/h2*2-4,8H,1H3,(H,9,10,11);;;1H2/q;;2*+1;/p-2
InChI key
AHTXUEDLGSAAMV-UHFFFAOYSA-L
storage temp.
Store at 0-8 C
Purity
97.00%
Country of Origin
CN
Product Link
https://www.achemblock.com/y227884-potassium-guaiacolsulfonate-hemihydrate.html
Signal Word
Warning
Hazard Statements
Precautionary statements
MSDS
https://www.achemblock.com/y227884-potassium-guaiacolsulfonate-hemihydrate.html
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
