Skip to Content
MilliporeSigma

ADVH9B9B4A8C

AChemBlock

SNC 80

Synonym(s):

4-[(R)-[(2S,5R)-4-allyl-2,5-dimethyl-1-piperazinyl](3-methoxyphenyl)methyl]-N,N-diethylbenzamide

Sign In to View Organizational & Contract Pricing.

Select a Size

Product ADVH9B9B4A8C is currently not available for purchase. Contact Technical Service

About This Item

Linear Formula:
C28H39O2N3
CAS Number:
Molecular Weight:
449.64
MDL number:

Skip To

Inchi Code

1S/C28H39N3O2/c1-7-17-30-19-22(5)31(20-21(30)4)27(25-11-10-12-26(18-25)33-6)23-13-15-24(16-14-23)28(32)29(8-2)9-3/h7,10-16,18,21-22,27H,1,8-9,17,19-20H2,2-6H3/t21-,22+,27-/m1/s1

Inchi Key

KQWVAUSXZDRQPZ-UMTXDNHDSA-N

Purity

97.00%

Storage temp.

Store at 0-8 C

Country of Origin

CN

Looking for similar products? Visit Product Comparison Guide

hstatements

signalword

Warning

pictograms

Exclamation mark

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library