ADVH9B9B4DE6
AChemBlock
IN-1130
Sign Into View Organizational & Contract Pricing
Select a Size
Product ADVH9B9B4DE6 is currently not available for purchase. Contact Technical Service
Skip To
Iupac Name
3-((4-(6-methylpyridin-2-yl)-5-(quinoxalin-6-yl)-1H-imidazol-2-yl)methyl)benzamide
Inchi Code
1S/C25H20N6O/c1-15-4-2-7-20(29-15)24-23(17-8-9-19-21(14-17)28-11-10-27-19)30-22(31-24)13-16-5-3-6-18(12-16)25(26)32/h2-12,14H,13H2,1H3,(H2,26,32)(H,30,31)
InChI key
RYKSGWSKILPDDY-UHFFFAOYSA-N
storage temp.
Store at 0-8 C
Purity
98%
Country of Origin
CN
Product Link
https://www.achemblock.com/z238311-in-1130.html
Signal Word
Warning
Hazard Statements
Precautionary statements
MSDS
https://www.achemblock.com/z238311-in-1130.html
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
