Skip to Content
MilliporeSigma
Get up to 22% off for Pi Day through March 27.Save Now

XPIH9BCF3C4D

AiFChem (an XtalPi company)

Methanesulfonic acid ((R)-1-((S)-2-amino-3-methylbutanoyl)pyrrolidin-2-yl)boronic acid (1:1)

Synonym(s):

methanesulfonic acid compound with (R)-1-((S)-2-amino-3-methylbutanoyl)pyrrolidin-2-ylboronic acid (1:1)

Sign In to View Organizational & Contract Pricing.

Select a Size


About This Item

Linear Formula:
C10H23O6N2S1B1
CAS Number:
Molecular Weight:
310.18
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

Inchi Code

1S/C9H19BN2O3.CH4O3S/c1-6(2)8(11)9(13)12-5-3-4-7(12)10(14)15;1-5(2,3)4/h6-8,14-15H,3-5,11H2,1-2H3;1H3,(H,2,3,4)/t7-,8-;/m0./s1

Inchi Key

OXYYOEIGQRXGPI-WSZWBAFRSA-N

Shipping Temperature

Keep container tightly closed in a dry and well-ventilated area. Containers which are opened must be carefully resealed and kept upright to prevent leakage.Keep in dark place,Inert atmosphere,2-8C

Purity

98%

Storage temp.

Keep container tightly closed in a dry and well-ventilated area. Containers which are opened must be carefully resealed and kept upright to prevent leakage.Keep in dark place,Inert atmosphere,2-8C

Country of Origin

CN

Physical Form

solid

Looking for similar products? Visit Product Comparison Guide


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service