Skip to Content
MilliporeSigma
Get 22% off for Pi Day through March 31.Save Now

XPIH9BCF96FD

AiFChem (an XtalPi company)

3,4-Dihydroxybenzaldehyde

Synonym(s):

3,4-dihydroxybenzaldehyde

Sign In to View Organizational & Contract Pricing.

Select a Size


About This Item

Linear Formula:
C7H6O3
CAS Number:
Molecular Weight:
138.12
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

Purity

98%

Inchi Code

1S/C7H6O3/c8-4-5-1-2-6(9)7(10)3-5/h1-4,9-10H

Inchi Key

IBGBGRVKPALMCQ-UHFFFAOYSA-N

Shipping Temperature

Keep container tightly closed in a dry and well-ventilated area. Containers which are opened must be carefully resealed and kept upright to prevent leakage.Keep in dark place,Sealed in dry,Room Temperature

Storage temp.

Keep container tightly closed in a dry and well-ventilated area. Containers which are opened must be carefully resealed and kept upright to prevent leakage.Keep in dark place,Sealed in dry,Room Temperature

Country of Origin

CN

Physical Form

Off-white to yellow to brown or pale-red or gray powder or

Looking for similar products? Visit Product Comparison Guide


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service