Skip to Content
MilliporeSigma

XPIH9BCFAA08

AiFChem (an XtalPi company)

4,6-Dichloro-2-methylquinoline

Synonym(s):

4,6-dichloro-2-methylquinoline

Sign In to View Organizational & Contract Pricing.

Select a Size

1 G

$68.00

5 G

$236.00

10 G

$393.00

Vendor SKU: ACGNMC201-1G

$68.00


Ships in 7 days - from Aldrich Partner.


About This Item

Linear Formula:
C10H7N1Cl2
CAS Number:
Molecular Weight:
212.08
MDL number:

Skip To

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

Country of Origin

CN

Physical Form

Form: solid

Inchi Code

1S/C10H7Cl2N/c1-6-4-9(12)8-5-7(11)2-3-10(8)13-6/h2-5H,1H3

Inchi Key

KYBCQZHRJZJYCE-UHFFFAOYSA-N

Shipping Temperature

Keep container tightly closed in a dry and well-ventilated area. Containers which are opened must be carefully resealed and kept upright to prevent leakage.Inert atmosphere,2-8C

Purity

98%

Storage temp.

Keep container tightly closed in a dry and well-ventilated area. Containers which are opened must be carefully resealed and kept upright to prevent leakage.Inert atmosphere,2-8C

Looking for similar products? Visit Product Comparison Guide


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service